Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004279.4 | XP_004004328.1 | 2 - 3 | 2165 | 104 | |
XM_012121475.3 | XP_011976865.1 | 2 - 3 | 2165 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |