Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010190.4 | XP_004010239.1 | 3 - 4 | 588 | 57 | |
XM_012180649.3 | XP_012036039.1 | 3 - 4 | 574 | 57 | ||
XM_012180650.3 | XP_012036040.1 | 3 - 4 | 758 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |