Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975418.1 | XP_027831219.1 | 3 - 4 | 539 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |