Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979096.1 | XP_027834897.1 | 10 - 11 | 5035 | 78 | |
XM_027979097.1 | XP_027834898.1 | 9 - 10 | 5406 | 78 | ||
XM_027979099.1 | XP_027834900.1 | 9 - 10 | 4864 | 78 | ||
XM_027979100.1 | XP_027834901.1 | 9 - 10 | 4636 | 78 | ||
XM_027979103.1 | XP_027834904.1 | 8 - 9 | 4465 | 78 | ||
XM_027979104.1 | XP_027834905.1 | 8 - 9 | 2791 | 78 | ||
XM_027979107.1 | XP_027834908.1 | 6 - 7 | 2221 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |