Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011337.4 | XP_004011386.1 | 4 - 5 | 836 | 68 | |
XM_004011338.4 | XP_004011387.1 | 4 - 5 | 836 | 68 | ||
XM_015097417.2 | XP_014952903.1 | 4 - 5 | 836 | 68 |
Gene Symbol: LOC114116100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972471.1 | XP_027828272.1 | 4 - 5 | 836 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |