Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013682.4 | XP_004013731.2 | 3 - 4 | 691 | 85 | |
XM_012187256.3 | XP_012042646.2 | 4 - 5 | 751 | 85 |
Gene Symbol: LOC114117224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975622.1 | XP_027831423.1 | 3 - 4 | 706 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |