Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100076.2 | XP_014955562.2 | 2 - 3 | 723 | 60 | |
XM_027977521.1 | XP_027833322.1 | 1 - 2 | 1011 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |