Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020464.4 | XP_004020513.2 | 3 - 4 | 727 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Ovis aries |