Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020017.4 | XP_004020066.1 | 5 - 6 | 698 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |