Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012174248.2 | XP_012029638.2 | 2 - 3 | 219 | 74 | |
XM_015094654.2 | XP_014950140.2 | 4 - 5 | 1585 | 74 | ||
XM_027967299.1 | XP_027823100.1 | 5 - 6 | 1187 | 74 | ||
XM_027967300.1 | XP_027823101.1 | 4 - 5 | 1529 | 74 |