Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096576.2 | XP_014952062.2 | 5 - 6 | 892 | 68 | |
XM_027971128.1 | XP_027826929.1 | 6 - 7 | 1003 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |