Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012129326.2 | XP_011984716.2 | 10 - 11 | 1339 | 68 | |
XM_012129337.2 | XP_011984727.1 | 10 - 11 | 1339 | 68 | ||
XM_012129346.2 | XP_011984736.1 | 10 - 11 | 1339 | 68 | ||
XM_027964623.1 | XP_027820424.1 | 9 - 10 | 1230 | 68 | ||
XM_027964624.1 | XP_027820425.1 | 9 - 10 | 1309 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |