Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004935.4 | XP_004004984.3 | 3 - 4 | 751 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |