Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012095631.2 | XP_011951021.2 | 2 - 3 | 219 | 112 | |
XM_012095634.2 | XP_011951024.2 | 2 - 3 | 233 | 112 |
Gene Symbol: LOC114111203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962888.1 | XP_027818689.1 | 1 | 161 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |