Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008938.4 | XP_004008987.1 | 3 - 4 | 265 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |