Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002672.3 | XP_004002721.2 | 7 - 8 | 974 | 75 | |
XM_027977088.1 | XP_027832889.1 | 7 - 8 | 1028 | 75 | ||
XM_027977095.1 | XP_027832896.1 | 7 - 8 | 1025 | 75 | ||
XM_027977099.1 | XP_027832900.1 | 7 - 8 | 986 | 75 | ||
XM_027977108.1 | XP_027832909.1 | 7 - 8 | 971 | 75 | ||
XM_027977116.1 | XP_027832917.1 | 7 - 8 | 932 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |