Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1H1orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001860.4 | XP_004001909.2 | 2 - 3 | 227 | 59 | |
XM_012112867.3 | XP_011968257.2 | 2 - 3 | 214 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |