Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006545.4 | XP_004006594.2 | 1 - 2 | 315 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |