Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002328.4 | XP_004002377.1 | 2 - 3 | 657 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |