Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099408.2 | XP_014954894.1 | 3 - 4 | 648 | 63 | |
XM_027976014.1 | XP_027831815.1 | 3 - 4 | 303 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |