Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963080.1 | XP_027818881.1 | 4 - 5 | 2710 | 70 | |
XM_027963081.1 | XP_027818882.1 | 5 - 6 | 2851 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |