Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969845.1 | XP_027825646.1 | 5 - 6 | 3450 | 88 | |
XM_027969846.1 | XP_027825647.1 | 4 - 5 | 3474 | 88 | ||
XM_027969847.1 | XP_027825648.1 | 6 - 7 | 3351 | 88 |