Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979327.1 | XP_027835128.1 | 2 - 3 | 920 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |