Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017067.3 | XP_004017116.2 | 2 - 3 | 156 | 73 |
Gene Symbol: LOC114113228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965211.1 | XP_027821012.1 | 1 | 110 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |