Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015091726.2 | XP_014947212.2 | 3 - 4 | 1994 | 92 | |
XM_027978489.1 | XP_027834290.1 | 4 - 5 | 1894 | 92 | ||
XM_027978499.1 | XP_027834300.1 | 4 - 5 | 1973 | 92 |