Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017183.3 | XP_004017232.3 | 5 - 6 | 1894 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |