Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959652.1 | XP_027815453.1 | 4 - 5 | 245 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |