Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012102042.2 | XP_011957432.1 | 6 - 7 | 982 | 77 | |
XM_012102044.3 | XP_011957434.1 | 6 - 7 | 997 | 77 | ||
XM_012102045.3 | XP_011957435.1 | 6 - 7 | 1016 | 77 | ||
XM_012102047.2 | XP_011957437.1 | 6 - 7 | 1351 | 77 | ||
XM_012102048.3 | XP_011957438.1 | 5 - 6 | 750 | 77 | ||
XM_012102049.3 | XP_011957439.1 | 5 - 6 | 842 | 77 | ||
XM_027960097.1 | XP_027815898.1 | 6 - 7 | 980 | 77 | ||
XM_027960098.1 | XP_027815899.1 | 6 - 7 | 980 | 77 | ||
XM_027960099.1 | XP_027815900.1 | 6 - 7 | 980 | 77 |