Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973525.1 | XP_027829326.1 | 3 - 4 | 596 | 94 | |
GenBank mRNA | EF121765.1 | - | 2 - 3 | 214 | 94 |