Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964478.1 | XP_027820279.1 | 1 - 2 | 165 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |