Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019613.4 | XP_004019662.1 | 1 - 2 | 1436 | 86 | |
XM_012102051.3 | XP_011957441.1 | 2 - 3 | 1012 | 86 |
Gene Symbol: LOC114117887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977913.1 | XP_027833714.1 | 1 | 300 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |