Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004397.3 | XP_004004446.2 | 4 - 5 | 671 | 226 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |