Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964060.1 | XP_027819861.1 | 5 - 6 | 1522 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |