Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002305.4 | XP_004002354.3 | 8 - 9 | 1530 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |