Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012580.4 | XP_004012629.2 | 5 - 6 | 785 | 101 | |
XM_027975103.1 | XP_027830904.1 | 4 - 5 | 646 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |