Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018797.4 | XP_004018846.3 | 1 - 2 | 504 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |