Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973352.1 | XP_027829153.1 | 2 - 3 | 2576 | 62 | |
XM_027973355.1 | XP_027829156.1 | 2 - 3 | 2576 | 62 | ||
XM_027973356.1 | XP_027829157.1 | 3 - 4 | 574 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |