Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960789.1 | XP_027816590.1 | 6 - 7 | 785 | 60 | |
XM_027960790.1 | XP_027816591.1 | 4 - 5 | 524 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |