Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106372.3 | XP_011961762.1 | 11 - 12 | 2203 | 97 | |
XM_015104819.2 | XP_014960305.1 | 11 - 12 | 2188 | 97 | ||
XM_015104820.2 | XP_014960306.1 | 10 - 11 | 1996 | 97 | ||
XM_015104821.2 | XP_014960307.1 | 11 - 12 | 2188 | 97 | ||
XM_015104822.2 | XP_014960308.1 | 11 - 12 | 2128 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |