Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106372.3 | XP_011961762.1 | 6 - 7 | 1698 | 105 | |
XM_015104819.2 | XP_014960305.1 | 6 - 7 | 1683 | 105 | ||
XM_015104820.2 | XP_014960306.1 | 5 - 6 | 1491 | 105 | ||
XM_015104821.2 | XP_014960307.1 | 6 - 7 | 1683 | 105 | ||
XM_015104822.2 | XP_014960308.1 | 6 - 7 | 1623 | 105 | ||
XM_015104824.2 | XP_014960310.1 | 6 - 7 | 1683 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |