Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022151.4 | XP_004022200.1 | 1 - 2 | 738 | 88 | |
XM_012106862.3 | XP_011962252.1 | 1 - 2 | 670 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |