Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC11C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103798.2 | XP_014959284.2 | 2 - 3 | 292 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |