Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012180288.3 | XP_012035678.1 | 9 - 10 | 1237 | 109 | |
XM_027971150.1 | XP_027826951.1 | 9 - 10 | 1312 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |