Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006187.4 | XP_004006236.2 | 7 - 8 | 1230 | 82 | |
XM_012173910.2 | XP_012029300.2 | 6 - 7 | 871 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |