Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013834.4 | XP_004013883.1 | 5 - 6 | 1932 | 79 | |
XM_012187624.3 | XP_012043014.1 | 4 - 5 | 1869 | 79 | ||
XM_012187626.3 | XP_012043016.1 | 5 - 6 | 1932 | 79 | ||
XM_012187627.3 | XP_012043017.1 | 4 - 5 | 1869 | 79 | ||
XM_027975915.1 | XP_027831716.1 | 5 - 6 | 963 | 79 | ||
XM_027975916.1 | XP_027831717.1 | 5 - 6 | 963 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |