Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959682.1 | XP_027815483.1 | 4 - 5 | 778 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |