Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003543.3 | XP_004003592.2 | 4 - 5 | 601 | 83 | |
XM_012149371.2 | XP_012004761.2 | 3 - 4 | 469 | 83 | ||
XM_027970500.1 | XP_027826301.1 | 2 - 3 | 492 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |