Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008465.4 | XP_004008514.1 | 5 - 6 | 746 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |