Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017250.4 | XP_004017299.1 | 2 - 3 | 2997 | 90 | |
XM_012097478.3 | XP_011952868.1 | 2 - 3 | 2994 | 90 | ||
XM_027956274.1 | XP_027812075.1 | 3 - 4 | 2742 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |