Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101121723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019389.4 | XP_004019438.2 | 4 - 5 | 648 | 92 | |
XM_012101586.3 | XP_011956976.2 | 4 - 5 | 627 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |